Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AQUA™ Peptide GFGFV[K], MilliporeSigma™ Supelco™

Catalog No. 111004162 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 pmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4162 100 pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4162 Supplier MilliporeSigma Supelco Supplier No. AQ0322100PMOL

Specifications

Product Line AQUA
Shipping Condition Wet Ice
Linear Formula C2713C6H47N515N2O7
Type Standardization Solution
Quantity 100 pmol
Formula Weight 661.71

This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School. Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.