Learn More
Polysciences, Inc. Aqua-Poly/Mount 20ml A water-soluble non-fluorescing mounting medium Useful for immunofluorescent techniques as it enhances and retains fluorescent stains Handling: Gloves and chemical goggles Storage: Store at 4 degrees celsius
SDP
Catalog No. NC9439247
Encompass_Preferred
Aqua-Poly/Mount 20ML
Catalog No. NC9439247 Supplier Polysciences, Inc. Supplier No. 1860620
Aqua-Poly/Mount 20ML