Learn More
Polysciences, Inc. Aqua-Poly/Mount, 5 x 20 mL
SDP
Catalog No. NC0128658
Encompass_Preferred
Aqua-Poly/Mount is a water-soluble, non-fluorescing, non-permanent, aqueous mounting medium formulated for mounting sections directly from aqueous solutions. It is useful for immunofluorescent techniques as it enhances and retains fluorescent stains.
Catalog No. NC0128658 Supplier Polysciences, Inc. Supplier No. 186065
Aqua-Poly/Mount is a water-soluble, non-fluorescing, non-permanent, aqueous mounting medium formulated for mounting sections directly from aqueous solutions. It is useful for immunofluorescent techniques as it enhances and retains fluorescent stains.