Learn More
Aqua Solutions New Mixed TC/TN Standard 2 (TOC 100ppm, IC 25ppm, TC 125ppm, TN 30ppm) | 250mL
SDP
Catalog No. NC1817649
Encompass_Preferred
New Mixed TC/TN Standard 2 (TOC 100ppm, IC 25ppm, TC 125ppm, TN 30ppm) | 250mL
Catalog No. NC1817649 Supplier Aqua Solutions Supplier No. SPX519
New Mixed TC/TN Standard 2 (TOC 100ppm, IC 25ppm, TC 125ppm, TN 30ppm) | 250mL