Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Aqua Solutions Acetone Reagent, ACS Reagent, 4 L, PVC Coated Glass
SDP

Catalog No. NC3994849
Encompass_Preferred

This acetone is an ACS Reagent grade chemical supplied in a 4 L Polyvinyl Chloride (PVC) coated glass container, suitable for laboratory applications.

  • Grade: ACS Reagent
  • CAS number: 67-64-1
  • Formula weight: 58.08
  • Volume: 4 L
  • Container type: Polyvinyl chloride (PVC) coated glass
  • Hazmat item
  • Packing group: II
  • UN number: UN1090

Catalog No. NC3994849 Supplier Aqua Solutions Supplier No. A14944LP
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This acetone is an ACS Reagent grade chemical supplied in a 4 L Polyvinyl Chloride (PVC) coated glass container, suitable for laboratory applications.

  • Grade: ACS Reagent
  • CAS number: 67-64-1
  • Formula weight: 58.08
  • Volume: 4 L
  • Container type: Polyvinyl chloride (PVC) coated glass
  • Hazmat item
  • Packing group: II
  • UN number: UN1090

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.