Learn More
Aqua Solutions Aqua Solutions Hydroxylamine Hydrochloride, 300 G/L, 500 mL
SDP
Catalog No. NC3994865
Encompass_Preferred
This Hydroxylamine Hydrochloride solution is formulated with a concentration of 300 G/L and provided in a 500 mL volume. It adheres to the ASTM E-200 assay standard, making it suitable for laboratory applications requiring precise chemical specifications. Classified as a hazardous and corrosive liquid, it requires careful handling.
  • Formulated with a concentration of 300 G/L
  • Supplied in a 500 mL volume
  • Conforms to ASTM E-200 assay standards
  • Classified as a hazardous material
  • Identified as a corrosive liquid
  • Features a packing group of III
  • Assigned UN number UN1760
Catalog No. NC3994865 Supplier Aqua Solutions Supplier No. 4460500ML
This Hydroxylamine Hydrochloride solution is formulated with a concentration of 300 G/L and provided in a 500 mL volume. It adheres to the ASTM E-200 assay standard, making it suitable for laboratory applications requiring precise chemical specifications. Classified as a hazardous and corrosive liquid, it requires careful handling.
  • Formulated with a concentration of 300 G/L
  • Supplied in a 500 mL volume
  • Conforms to ASTM E-200 assay standards
  • Classified as a hazardous material
  • Identified as a corrosive liquid
  • Features a packing group of III
  • Assigned UN number UN1760