Learn More
Adam Equipment Adam Equipment Aqua Stainless Steel Washdown Scale ABW 16S 35lb / 16kg capacity x 0.002lb / 1g
Supplier:  Adam Equipment ABW 16S
Encompass_Preferred
Adam Equipment Aqua Washdown Scales boasts IP68 washdown protection, meaning it's suitable for weighing materials in demanding environments. In addition to the easy-to-clean grade 304 stainless steel housing and platform, the pan is removable for hygienic maintenance. Overload protection helps safeguard the scale against accidental overloads for a reliable, long-lasting weighing solution. Well-suited for food processing, batching and packaging, designed to improve productivity. The bright LED displays clearly shows results in the front and rear of the scale, allowing multiple users to use the same scale, while hidden color-changing indicators light up during checkweighing to show if results are over, under or within preset limits. Percentage weighing makes baking and recipe batching effortless, while parts counting speeds up packaging tasks. Internal rechargeable battery for operation anywhere. Rechargable Battery & AC Adapter included
Catalog No. NC2612385