Learn More
Adam Equipment Adam Equipment Aqua Washdown Scale ABW 4 9lb / 4kg capacity x 0.001lb / 0.0005kg readability
Catalog No. NC2612399
Encompass_Preferred
Adam Equipment ABW Washdown Scales are ideal for food preparation, food processing units and harsh industrial environments, Aqua washdown scales are highly versatile. They're rated IP67 and feature a large removable stainless steel pan, so they're protect
Catalog No. NC2612399 Supplier Adam Equipment Supplier No. ABW 4
Adam Equipment ABW Washdown Scales are ideal for food preparation, food processing units and harsh industrial environments, Aqua washdown scales are highly versatile. They're rated IP67 and feature a large removable stainless steel pan, so they're protect