Learn More
Adam Equipment Adam Equipment Aqua Washdown Scale ABW 32 70lb / 32kg capacity x 0.01lb / 5g readability
Catalog No. NC2612400
Encompass_Preferred
Adam Equipment ABW Washdown Scales are ideal for food preparation, food processing units and harsh industrial environments, Aqua washdown scales are highly versatile. They're rated IP67 and feature a large removable stainless steel pan, so they're protect
Catalog No. NC2612400 Supplier Adam Equipment Supplier No. ABW 32
Adam Equipment ABW Washdown Scales are ideal for food preparation, food processing units and harsh industrial environments, Aqua washdown scales are highly versatile. They're rated IP67 and feature a large removable stainless steel pan, so they're protect