Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation AquaAnalysis System Start-up Solution, Liquid, 2 mL, 12 months Shelf Life

Catalog No. 50201680 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

  • AquaAnalysis System standards kit for system assessment.
  • Contains 1 ampule of 1.0 mL organophosphorus mix.
  • Dicrotophos 1.00 mg/mL ± 0.5%.
  • Malathion 1.00 mg/mL ± 0.5%.
  • Solvent: Acetonitrile.
  • Separation type: Reversed phase.
  • Detection method: MS.
  • Formulation: Liquid.
  • Storage condition: Room temperature.
  • System type: MS.
  • Device: AquaAnalysis System.
  • Shelf life: 12 months.
  • Shipping condition: Ambient.
  • Volume: 2 mL.
  • Type: Installation.
  • Application: System performance.
  • Product type: Standards.
  • Units per package: 1.
  • Includes Certificate of Analysis.

Catalog No. 50-201-680 Supplier Waters Corp Supplier No. 700003870
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • AquaAnalysis System standards kit for system assessment.
  • Contains 1 ampule of 1.0 mL organophosphorus mix.
  • Dicrotophos 1.00 mg/mL ± 0.5%.
  • Malathion 1.00 mg/mL ± 0.5%.
  • Solvent: Acetonitrile.
  • Separation type: Reversed phase.
  • Detection method: MS.
  • Formulation: Liquid.
  • Storage condition: Room temperature.
  • System type: MS.
  • Device: AquaAnalysis System.
  • Shelf life: 12 months.
  • Shipping condition: Ambient.
  • Volume: 2 mL.
  • Type: Installation.
  • Application: System performance.
  • Product type: Standards.
  • Units per package: 1.
  • Includes Certificate of Analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.