Learn More
Description
Thermo Scientific™ Aquabath Water Baths provide precise temperature control and are available with an analog or digital operating system. The Aquabath is available in a variety of sizes from 2 to 28L, including a shallow form to meet specific application needs and budgets. Bath features dual thermostats and an independent high-limit thermostat for overtemperature protection.
- Hinged acrylic cover lifts to a 90° stay-open position or can be removed completely
- “Fins” on the hinged clear acrylic gable cover (included) protect hands from hot vapors
- Exterior remains cool to the touch even after extended use to prevent accidental burns
- Temperature-resistant plastic rim fits over the seamless stainless-steel reservoir for a leak-free seal
- Stainless-steel exterior resists corrosion and has a tough, easy-to-clean enamel coating
- Rounded, seamless stainless-steel reservoir and diffuser tray resist rust and contamination for simple maintenance
- Movable temperature controllers on the 20 and 28L digital models can be used on the bath long or short side to best utilize bench space
- Heating elements will not burn out if the bath accidentally runs dry
- Drain pump is included with 10, 20, 28 and 5/10L models
Digital Operating System
- Field-calibratable
- PID control provides ±0.24°C uniformity, ±0.5°C stability and ±0.1°C control resolution at 37°C
- Digital temperature set and readout selectable display with 0.1° resolution
Analog Operating System
- Easy-to-use analog control and hydraulic thermostat—ideal for fixed-setpoint applications
- Temperature uniformity ±0.2°C at 37°C
- Diffuser tray, thermometer clip and nonhazardous liquid, partial immersion (76mm) thermometer (-10° to +110°C) included (230V models do not include a thermometer)
Ordering Information:
Stainless-steel covers available separately.
Compatible with:
- Cover, for 2L baths, stainless steel (19000-11Q)
- Cover, for 2L shallow and 5L baths, stainless steel (19000-13Q)
- Cover, for 20L and 28L baths, stainless steel (19000-17Q)
- Cover, for 10L baths, stainless steel (19000-15Q)
Specifications
Specifications
| Chamber Material | Stainless Steel |
| Controller Type | Digital |
| Capacity (Metric) | 20 L |
| Certifications/Compliance | CSA and CE certified |
| Pump Type | Drain Pump |
| Temperature Range | Ambient to 100°C |
| Material | Enamel-coated Stainless Steel Shell |
| Temperature Uniformity | ±0.24° at 37°C |
| Dimensions (L x W x H) | 16.4 x 24.2 x 8.9 in. (42 x 62 x 23 cm) |
| Length (English) | 16.4 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.