Learn More
AQUABILITI Aquabiliti Bacteriostatic Water, 0.9% Benzyl Alcohol, USP Grade, 1100 L
Catalog No. NC3947900
Encompass_Preferred
This bulk packaged Bacteriostatic Water consists of ultra-high-purity Water for Injection (WFI) supplemented with 0.9% Benzyl Alcohol as a preservative. Designed for further manufacturing applications, this product is supplied in customizable single-use bioprocess containers (BPCs) suitable for large-scale drug production and repackaging.
  • Manufactured in accordance with cGMP standards
  • Meets strict USP guidelines for quality and consistency
  • Available in scalable single-use formats
  • Formulated with 0.9% benzyl alcohol preservative
  • Utilizes ultra-high-purity water for injection base
Catalog No. NC3947900 Supplier AQUABILITI Supplier No. BAC110001
This bulk packaged Bacteriostatic Water consists of ultra-high-purity Water for Injection (WFI) supplemented with 0.9% Benzyl Alcohol as a preservative. Designed for further manufacturing applications, this product is supplied in customizable single-use bioprocess containers (BPCs) suitable for large-scale drug production and repackaging.
  • Manufactured in accordance with cGMP standards
  • Meets strict USP guidelines for quality and consistency
  • Available in scalable single-use formats
  • Formulated with 0.9% benzyl alcohol preservative
  • Utilizes ultra-high-purity water for injection base