Learn More
AQUABILITI Aquabiliti Purified Water, Sterile, USP Grade, 20 L
Catalog No. NC3947884
Encompass_Preferred
Sterile purified water manufactured and tested in accordance with the USP monograph for sterile purified water. It is suitable for both sterile and non-sterile applications, including use in oral and topical products, non-active medical device manufacturing, and various cleaning processes.
  • Complies with USP monograph standards
  • Suitable for non-parenteral applications
  • Sterility tested with no growth
  • Total organic carbon level below 8000 ppb
  • Conductivity less than 5 µS/cm
  • Clear formulation
  • Provided in a 20L pour bag
Catalog No. NC3947884 Supplier AQUABILITI Supplier No. SPW002001
Sterile purified water manufactured and tested in accordance with the USP monograph for sterile purified water. It is suitable for both sterile and non-sterile applications, including use in oral and topical products, non-active medical device manufacturing, and various cleaning processes.
  • Complies with USP monograph standards
  • Suitable for non-parenteral applications
  • Sterility tested with no growth
  • Total organic carbon level below 8000 ppb
  • Conductivity less than 5 µS/cm
  • Clear formulation
  • Provided in a 20L pour bag