Learn More
AQUABILITI Aquabiliti Purified Water, Sterile, USP Grade, 20 L
Catalog No. NC3947885
Encompass_Preferred
This sterile, non-pyrogenic purified water is manufactured and tested in strict accordance with the USP monograph for sterile purified water. It is formulated for use in a variety of non-parenteral applications, including the manufacture and cleaning of non-active medical devices, as well as in the production of oral and topical products.
  • Manufactured and tested per USP monograph standards
  • Sterile and non-pyrogenic formulation
  • Suitable for non-active medical device manufacturing and cleaning
  • Appropriate for use in oral and topical products
  • For further manufacturing use only; not for direct therapeutic use
Catalog No. NC3947885 Supplier AQUABILITI Supplier No. SPW002002
This sterile, non-pyrogenic purified water is manufactured and tested in strict accordance with the USP monograph for sterile purified water. It is formulated for use in a variety of non-parenteral applications, including the manufacture and cleaning of non-active medical devices, as well as in the production of oral and topical products.
  • Manufactured and tested per USP monograph standards
  • Sterile and non-pyrogenic formulation
  • Suitable for non-active medical device manufacturing and cleaning
  • Appropriate for use in oral and topical products
  • For further manufacturing use only; not for direct therapeutic use