Learn More
AQUABILITI Aquabiliti Water For Injection, USP, EP, Clear, 10 L
Catalog No. NC3947917
Encompass_Preferred
This multi-compendial Water for Injection (WFI) quality water is manufactured to meet rigorous United States Pharmacopeia (USP) and European Pharmacopoeia (EP) standards. Packaged in bulk for commercial use, this product is suitable for upstream and downstream bioprocessing and general manufacturing applications. It is intended for further manufacturing use only and is not for direct therapeutic use.
  • Complies with USP/EP monographs for WFI quality water
  • Endotoxin limit ≤ 0.25 EU/mL
  • Total organic carbon packaged ≤ 8.00 mg/L
  • Conductivity packaged ≤ 5 µS/cm
  • Particulate matter specification ≤ 3 particles/mL
  • Validated sterile with no growth
  • Clear and colorless appearance
Catalog No. NC3947917 Supplier AQUABILITI Supplier No. HYPER001003
This multi-compendial Water for Injection (WFI) quality water is manufactured to meet rigorous United States Pharmacopeia (USP) and European Pharmacopoeia (EP) standards. Packaged in bulk for commercial use, this product is suitable for upstream and downstream bioprocessing and general manufacturing applications. It is intended for further manufacturing use only and is not for direct therapeutic use.
  • Complies with USP/EP monographs for WFI quality water
  • Endotoxin limit ≤ 0.25 EU/mL
  • Total organic carbon packaged ≤ 8.00 mg/L
  • Conductivity packaged ≤ 5 µS/cm
  • Particulate matter specification ≤ 3 particles/mL
  • Validated sterile with no growth
  • Clear and colorless appearance