Learn More
AQUABILITI Aquabiliti Water For Injection, USP/EP Grade, Clear, Bioprocess 3D Container, 50 L
Catalog No. NC3947821
Encompass_Preferred
AQUAPure HYPER Water for Injection (WFI) is a high-purity, USP/EP grade water solution supplied in a 50L bioprocess 3D container (BPC). Designed for demanding bioprocessing environments, this water meets strict pharmacopeia standards for use in upstream and downstream manufacturing applications.
  • Meets USP and EP quality standards for water for injection
  • Supplied in a 50L bioprocess 3D container for sterile transfer
  • Controlled endotoxin levels of 0.05 EU/mL
  • Complies with USP <643> for total organic carbon
  • Complies with USP <645> for conductivity
  • Sterility tested per USP <71> with no growth observed
  • Particulate matter verified per USP <788>
Catalog No. NC3947821 Supplier AQUABILITI Supplier No. HYPER005001
AQUAPure HYPER Water for Injection (WFI) is a high-purity, USP/EP grade water solution supplied in a 50L bioprocess 3D container (BPC). Designed for demanding bioprocessing environments, this water meets strict pharmacopeia standards for use in upstream and downstream manufacturing applications.
  • Meets USP and EP quality standards for water for injection
  • Supplied in a 50L bioprocess 3D container for sterile transfer
  • Controlled endotoxin levels of 0.05 EU/mL
  • Complies with USP <643> for total organic carbon
  • Complies with USP <645> for conductivity
  • Sterility tested per USP <71> with no growth observed
  • Particulate matter verified per USP <788>