Learn More
AQUABILITI Aquabiliti Water For Injection, USP/EP Grade, PETG, 20 L
Catalog No. NC3947822
Encompass_Preferred
This multi-compendial Water for Injection (WFI) quality water is designed for upstream and downstream bioprocessing and manufacturing applications. It is produced through a multi-stage purification system featuring reverse osmosis, electrodeionization, ultraviolet oxidation, and final polishing. The product is provided in a 20L bioprocess container system that utilizes single-use components to minimize contamination risks.
  • Meets USP and EP quality standards
  • Packaged in 20L recyclable PETG trays and bioprocess containers
  • Features invertible lid design for complete fluid evacuation
  • Includes sterile quick connectors and integrated 3/8-inch wall tubing
  • Monitored against USP <643>, <645>, <85>, <71>, and <788> testing parameters
  • Eliminates cross-contamination with a closed-loop, single-use system
Catalog No. NC3947822 Supplier AQUABILITI Supplier No. HYPER002001
This multi-compendial Water for Injection (WFI) quality water is designed for upstream and downstream bioprocessing and manufacturing applications. It is produced through a multi-stage purification system featuring reverse osmosis, electrodeionization, ultraviolet oxidation, and final polishing. The product is provided in a 20L bioprocess container system that utilizes single-use components to minimize contamination risks.
  • Meets USP and EP quality standards
  • Packaged in 20L recyclable PETG trays and bioprocess containers
  • Features invertible lid design for complete fluid evacuation
  • Includes sterile quick connectors and integrated 3/8-inch wall tubing
  • Monitored against USP <643>, <645>, <85>, <71>, and <788> testing parameters
  • Eliminates cross-contamination with a closed-loop, single-use system