Learn More
AQUABILITI Aquabiliti Water For Injection, USP Grade, Bioprocess 3D Container, 100 L
Catalog No. NC3947810
Encompass_Preferred
This product is a USP grade water for injection provided in a 100L bioprocess 3D container. It is specifically formulated for further manufacturing applications and is not intended for direct therapeutic use.
  • Clear and colorless liquid appearance
  • Total organic carbon level of 20 ppb
  • Conductivity measured at 0.06 µS/cm
  • Endotoxin concentration of 0.03 EU/mL
  • Validated for sterility with no growth observed
  • Particulate matter limit of 3 particles/mL
Catalog No. NC3947810 Supplier AQUABILITI Supplier No. ULTRA010001
This product is a USP grade water for injection provided in a 100L bioprocess 3D container. It is specifically formulated for further manufacturing applications and is not intended for direct therapeutic use.
  • Clear and colorless liquid appearance
  • Total organic carbon level of 20 ppb
  • Conductivity measured at 0.06 µS/cm
  • Endotoxin concentration of 0.03 EU/mL
  • Validated for sterility with no growth observed
  • Particulate matter limit of 3 particles/mL