Learn More
AQUABILITI Aquabiliti Water For Injection, USP Grade, Bioprocess 3D Container, 50 L
Catalog No. NC3947811
Encompass_Preferred
This product provides high-purity Water for Injection (WFI) quality water, meeting established USP grade monographs for commercial and industrial applications. It is supplied in a sterile 50L Bioprocess 3D Container (BPC) optimized for large-scale bioprocessing workflows.
  • Meets USP monograph standards for water for injection quality
  • Conductivity measured at 0.06 μS/cm
  • Total organic carbon (TOC) levels of 50 ppb
  • Endotoxin levels limited to 0.03 EU/mL
  • Supplied in a 50L bioprocess 3d container
  • Intended for further manufacturing use only, not for direct therapeutic applications
Catalog No. NC3947811 Supplier AQUABILITI Supplier No. ULTRA005001
This product provides high-purity Water for Injection (WFI) quality water, meeting established USP grade monographs for commercial and industrial applications. It is supplied in a sterile 50L Bioprocess 3D Container (BPC) optimized for large-scale bioprocessing workflows.
  • Meets USP monograph standards for water for injection quality
  • Conductivity measured at 0.06 μS/cm
  • Total organic carbon (TOC) levels of 50 ppb
  • Endotoxin levels limited to 0.03 EU/mL
  • Supplied in a 50L bioprocess 3d container
  • Intended for further manufacturing use only, not for direct therapeutic applications