Learn More
AQUABILITI Aquabiliti Water For Injection, USP Grade, Sterile, 20 L
Catalog No. NC3947813
Encompass_Preferred
This USP-grade Water for Injection (WFI) is a high-purity liquid designed for critical pharmaceutical, biopharmaceutical, and medical device manufacturing workflows. Produced via a multi-stage purification system that includes reverse osmosis, electrodeionization, ultraviolet oxidation, and final polishing, this water consistently exceeds United States Pharmacopeia standards for WFI.
  • Meets USP monograph specifications for water for injection
  • Processed through advanced multi-stage purification technology
  • Sterile composition to support high-integrity manufacturing
  • Integrated bag and tray system for contamination control
  • Invertible lid design for complete fluid evacuation
  • Thick-wall tubing suitable for peristaltic pump dispensing
  • Recyclable tray construction
  • Poly overwrap packaging to minimize particulate risk
Catalog No. NC3947813 Supplier AQUABILITI Supplier No. ULTRA002002
This USP-grade Water for Injection (WFI) is a high-purity liquid designed for critical pharmaceutical, biopharmaceutical, and medical device manufacturing workflows. Produced via a multi-stage purification system that includes reverse osmosis, electrodeionization, ultraviolet oxidation, and final polishing, this water consistently exceeds United States Pharmacopeia standards for WFI.
  • Meets USP monograph specifications for water for injection
  • Processed through advanced multi-stage purification technology
  • Sterile composition to support high-integrity manufacturing
  • Integrated bag and tray system for contamination control
  • Invertible lid design for complete fluid evacuation
  • Thick-wall tubing suitable for peristaltic pump dispensing
  • Recyclable tray construction
  • Poly overwrap packaging to minimize particulate risk