Learn More
AQUABILITI Aquabiliti Water For Irrigation, USP Grade, Sterile, 20 L
Catalog No. NC3947875
Encompass_Preferred
This is a sterile, non-pyrogenic water product manufactured and tested in compliance with the USP monograph for bulk packaged water for irrigation. Produced using a proprietary purification system, this water is suitable for industrial applications including the manufacturing and testing of medical devices, and serves as an alternative in processes where Water for Injection is not required. This product is for further manufacturing use only and is not intended for direct therapeutic applications.
  • Meets USP monograph specifications for water for irrigation
  • Sterile and non-pyrogenic liquid
  • Clear and colorless appearance
  • Supplied in a 20L bioprocess 2D container
  • Suitable for medical device manufacturing and testing
Catalog No. NC3947875 Supplier AQUABILITI Supplier No. WIR002002
This is a sterile, non-pyrogenic water product manufactured and tested in compliance with the USP monograph for bulk packaged water for irrigation. Produced using a proprietary purification system, this water is suitable for industrial applications including the manufacturing and testing of medical devices, and serves as an alternative in processes where Water for Injection is not required. This product is for further manufacturing use only and is not intended for direct therapeutic applications.
  • Meets USP monograph specifications for water for irrigation
  • Sterile and non-pyrogenic liquid
  • Clear and colorless appearance
  • Supplied in a 20L bioprocess 2D container
  • Suitable for medical device manufacturing and testing