Learn More
FUJIFILM Biosciences AquaBlot 10X Hgh Eff Trans Buf
Catalog No. 5000450311
Encompass_Preferred
AquaBlot 10X High Efficiency Transfer Buffer is a high-performance liquid buffer used for protein transfer in blotting applications This is a 10 transfer buffer superior in transfer efficiency to the Towbin buffer Tris-glycine-methanol Dilute 10 times with deionized water before use No methanol needs to be added Methanol is necessary for wetting a PVDF membrane Higher transfer efficiency than conventional methodsSimply change the transfer bufferNo methanol addition required For research use only RUO
Catalog No. 50-004-50311 Supplier FUJIFILM Biosciences Supplier No. 01526213
AquaBlot 10X High Efficiency Transfer Buffer is a high-performance liquid buffer used for protein transfer in blotting applications This is a 10 transfer buffer superior in transfer efficiency to the Towbin buffer Tris-glycine-methanol Dilute 10 times with deionized water before use No methanol needs to be added Methanol is necessary for wetting a PVDF membrane Higher transfer efficiency than conventional methodsSimply change the transfer bufferNo methanol addition required For research use only RUO