Learn More
Andwin Scientific Scienceware Cleanware Aqua-Clear Water Conditioner 100ml
SDP
Catalog No. 50999422
Encompass_Preferred
The SCIENCEWARE Aqua-Clear Water Conditioner prevent the growth of algae, bacteria, molds, and biofilms in water systems. This product does not harm film. It also stops clogging of tubings. This product is sold in a quantity of 100 mL.
Catalog No. 50-999-422 Supplier Andwin Scientific Supplier No. S10502
The SCIENCEWARE Aqua-Clear Water Conditioner prevent the growth of algae, bacteria, molds, and biofilms in water systems. This product does not harm film. It also stops clogging of tubings. This product is sold in a quantity of 100 mL.