Learn More
Jm Science Coulometric Karl Fischer Reagent, 1 L, pyridine-free, for moisture determination
Catalog No. NC0268951
Encompass
This technical overview describes a high-performance coulometric Karl Fischer vessel solution designed for precise moisture determination in scientific applications. This pyridine-free anolyte is formulated for accurate and reproducible results with rapid analysis times. It is compatible with a wide range of coulometric Karl Fischer titrators and cell configurations, ensuring consistency and reliability across various applications.
  • High-performance moisture analysis
  • Pyridine-free composition enhances safety
  • Optimized formulation for fast pre-titration
  • Stable endpoint for consistent results
  • Extended shelf life for long-term use
  • Compatible with all titrators and cell types
  • Can be used with or without a diaphragm
  • Contains methyl alcohol
  • Hazmat classified
  • Not DEA controlled
  • Package options available: 1 L and 4 x 4 L
Catalog No. NC0268951 Supplier Jm Science Supplier No. 1612
This technical overview describes a high-performance coulometric Karl Fischer vessel solution designed for precise moisture determination in scientific applications. This pyridine-free anolyte is formulated for accurate and reproducible results with rapid analysis times. It is compatible with a wide range of coulometric Karl Fischer titrators and cell configurations, ensuring consistency and reliability across various applications.
  • High-performance moisture analysis
  • Pyridine-free composition enhances safety
  • Optimized formulation for fast pre-titration
  • Stable endpoint for consistent results
  • Extended shelf life for long-term use
  • Compatible with all titrators and cell types
  • Can be used with or without a diaphragm
  • Contains methyl alcohol
  • Hazmat classified
  • Not DEA controlled
  • Package options available: 1 L and 4 x 4 L