Learn More
CTL Scientific Supply Corp AQUADUR,100 TEST STRIPS
SDP
Supplier:  CTL Scientific Supply Corp 91220
Encompass_Preferred
AQUADUR are test sticks for determination of the water hardness. When measuring the total hardness, the sum of these salts is determined. Gradation of scale: <55 - >70 - >125 - >250 - >375 ppm. Contains 100 test strips
Catalog No. 50-117-3316