Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqualine™ Complete 2, Fisher Chemical™

Catalog No. AL1950212 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass
Amber glass bottle
Color:
Colorless
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Color
AL-1950212 2.5 L Amber glass bottle Colorless
AL-19501 1 L Amber Glass Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AL-1950212
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Single-component reagent; Pyridine free

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 2.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aqualine™ Complete 2
Name Note Karl Fischer Reagent
Color Colorless
CAS 7446-09-5, 288-32-4, 111-90-0, 7553-56-2
Physical Form Liquid
Quantity 2.5 L
Molecular Formula Solution
Packaging Amber glass bottle
MDL Number 81734
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.