Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Catalog No. al20001 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
2.5 L
4 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Single-component reagent; Pyridine free

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 5.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Chemical Name or Material Aqualine™ Complete 5
Quantity 1 L
Purity or Free of Statement Pyridine free
Name Note Karl Fischer Reagent
CAS 111-90-0, 288-32-4, 7446-09-5, 7553-09-5
Equivalence 5.2mg of water/mL min.
Packaging Amber glass bottle
Physical Form Liquid
Molecular Formula C29H50O3
MDL Number 81734
InChI Key LTVDFSLWFKLJDQ-UHFFFAOYNA-N
Molecular Weight (g/mol) 446.72
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.