Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Aqualine™ Complete 5K, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Catalog No. p-305673 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AL2250R-1 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AL2250R-1
Only null left
Add to Cart
This item is not returnable. View return policy
  • Single-component volumetric reagent for high water content analysis of aldehydes and ketones
  • Water equivalent: 5.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aqualine™ Complete 5K
Name Note Karl Fischer Reagent
Color Colorless
CAS 7446-09-5, 5683-33-0, 111-90-0, 7553-56-2
Physical Form Liquid
Quantity 1 L
Packaging Amber glass bottle
MDL Number 81734
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.