Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Aqualine™ Solvent, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Catalog No. p-305695 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AL-21001 1 L
AL-2100212 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AL-21001
Only null left
Add to Cart
This item is not returnable. View return policy

Solvent for two-component system; Pyridine free

  • Used in conjunction with Aqualine™ titrant for two component volumetric Karl Fischer analysis
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aqualine™ Solvent
Name Note Karl Fischer Reagent
Color Colorless
CAS 7446-09-5, 288-32-4
Physical Form Liquid
Quantity 1 L
Packaging Amber glass bottle
MDL Number 81734
UN Number 1992
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.