Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Aqualine™ Water Standard 10mg, for Karl Fischer Titration, Fisher Chemical

Catalog No. AL272080 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 x 8 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AL-272080 10 x 8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AL-272080
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

10mg/mL H2O standard

  • Standard contains 10mg H2O/mL
  • Used for titer determination of volumetric Karl Fischer reagents
  • Supplied as 10 x 8mL and packaged in glass ampules to protect from moisture
  • The ampule should only be opened immediately prior to use to prevent absorption of moisture from atmosphere
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number 240423
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Physical Form Liquid
CAS 71-36-3
Quantity 10 x 8 mL
Packaging Ampoule
Chemical Name or Material Aqualine™ Standard 10.1, Karl Fischer Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.