Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Aqualine™ Titrant 5, for Karl Fischer Titration By Volumetry, Fisher Chemical

Catalog No. AL22001 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AL-22001 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AL-22001
Get Up to $1,800 in Chemicals with Purchase
View all promotions
 Restricted Item
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For two-component system. Water equivalent: 5.0mg H2O/mL

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aqualine™ Titrant 5
Name Note Karl Fischer Reagent
Boiling Point 64°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Molecular Formula CH4O
Packaging Amber glass bottle
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.