Learn More
Molecular Devices Molecular Devices Microplate Washer, 96/384-Well, 4-Channel, Touchscreen
Catalog No. 5000014690
Encompass_Preferred
The AquaMax 4000 Microplate Washer is a fully self-contained system for biochemical and cell-based microplate assays, configurable for both 96- and 384-well plates. It delivers precise dispense and aspiration control, interchangeable wash heads, and touchscreen operation to support reproducible workflows and gentle washing of weakly adherent cells.
  • Fully self-contained operation with onboard pumps and standby priming
  • Interchangeable 96- and 384-well wash heads for flexible workflows
  • Four source channels with level sensing for reagent bottles
  • Touchscreen interface with preconfigured and custom protocols
  • Compatible with plate stackers and robotic plate handlers for automation
  • Supports flat, U-, and V-bottom SBS-format microplates
  • Gentle washing options to minimize cell loss in cell-based assays
Catalog No. 50-000-14690 Supplier Molecular Devices Supplier No. AQUAMAX4KBUNDLE
Add to Cart
Add to Cart
Request a Quote
The AquaMax 4000 Microplate Washer is a fully self-contained system for biochemical and cell-based microplate assays, configurable for both 96- and 384-well plates. It delivers precise dispense and aspiration control, interchangeable wash heads, and touchscreen operation to support reproducible workflows and gentle washing of weakly adherent cells.
  • Fully self-contained operation with onboard pumps and standby priming
  • Interchangeable 96- and 384-well wash heads for flexible workflows
  • Four source channels with level sensing for reagent bottles
  • Touchscreen interface with preconfigured and custom protocols
  • Compatible with plate stackers and robotic plate handlers for automation
  • Supports flat, U-, and V-bottom SBS-format microplates
  • Gentle washing options to minimize cell loss in cell-based assays