Learn More
Molecular Devices Molecular Devices Automated Cell Culture System, AI-Driven, High Throughput
Supplier: Molecular Devices CELLXPRESSAI
The CellXpress.ai Automated Cell Culture System is an integrated, AI-driven platform that automates feeding, passaging, incubation, imaging, and analysis for 2D and 3D cell models, including stem cells, spheroids, and organoids. It enables continuous 24/7 operation to improve reproducibility, reduce manual error, and scale complex workflows for drug discovery and translational research.
- Automates feeding, passaging, and scaling of 2D and 3D cultures.
- Uses image-guided AI decision making to trigger culture events at the right time.
- Supports continuous 24/7 operation to increase throughput and reproducibility.
- Includes integrated incubation, high-content imaging, and liquid handling.
- Provides automated decontamination and HEPA-filtered enclosure for sterility.
- Enables high-throughput processing of over 100 plates in parallel.
- Delivers standardized, reproducible protocols and a complete experiment audit trail.