Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaMet 95.0+%, TCI America™
SDP

Catalog No. A3009100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3009100MG 100 mg
A3009500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3009100MG Supplier TCI America Supplier No. A3009100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1414707-08-6
Molecular Formula C39H55Cl3N4ORu
MDL Number MFCD28411647
Synonym [1,3-Bis(2,4,6-trimethylphenyl)-4-[(4-ethyl-4-methylpiperazinium-1-yl)methyl]-2-imidazolidinylidene]dichloro(2-isopropoxybenzylidene)ruthenium(VI) Chloride

Specifications

Color Green-Yellow
Quantity 100 mg
Formula Weight 803.32
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material AquaMet
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.