Learn More
Supply Solutions API Corp - Aquamicron Check Solution P. Size: 100ml; Rnage: 3.8 - 4.2mg water per ML.; Size: 100ml; Range: 3.8 - 4.2mg water per ML
Catalog No. NC0436627
Encompass
Aquamicron™ Check solution for coulometric Karl Fischer titration. 3.8 to 4.2 MG H2O/ML Methylcellusolve in 100 ML bottle.; Size: 100ml; Range: 3.8 - 4.2mg water per ML
Catalog No. NC0436627 Supplier Supply Solutions Supplier No. COSA MC02020
Aquamicron™ Check solution for coulometric Karl Fischer titration. 3.8 to 4.2 MG H2O/ML Methylcellusolve in 100 ML bottle.; Size: 100ml; Range: 3.8 - 4.2mg water per ML