Learn More
Thermo Scientific™ Aquanex™ DI Cartridge Set Package
Catalog No. 01670741 Shop All Thermo Scientific Products
Change view
Click to view available options
Feedwater Source:
Tap
Catalog No. Feedwater Source
01-670-741 Tap
1 options
Catalog No. 01-670-741 Supplier Thermo Scientific™ Supplier No. 7501050
Add to Cart
Add to Cart
The Thermo Scientific™ Aquanex™ DI Cartridge Packs simplify the cartridge installation process with a quick connect installation process. The Aquanex™ touchscreen will walk you through the consumable change process and log any consumable updates and recommended replacement dates.
The Thermo Scientific™ Aquanex™ is designed to truly help make life easier in lab with an intuitive touch screen interface, easy-access consumables, and smart dispensing features. The easy-to-use interface caters to users that need secured access and seamless event logging, system monitoring, and reporting. A new innovative cartridge design provides low TOC, a longer cartridge life, and an easy-to-change design.
Easily access consumables
  • The Aquanex user interface provides step-by-step instructions on how to access and change consumables
  • UV lamp, RO assembly, and ultrafilter consumables are easily accessed by removing the magnetic front panel
  • No screws or complicated tools are required to change consumables
Auto-detection and event logging
  • Aquanex system automatically detects and logs when the DI cartridges are replaced
  • The system will notify you when consumables are nearing end of life and require replacement
  • Consumable installation and replacement dates can be easily viewed in the maintenance menu
Simplified DI cartridge installation
  • No water spills or mess during cartridge installation or removal
  • Eject the DI cartridge by simply pressing a button on the side of the system
  • Guide rails, built-in sensors, and a quick-connect system allow for smooth installation of DI cartridges
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Tap
For Use With (Equipment) Aquanex Water Purification System
Includes A and B Replacement Cartridges