Learn More
Biotium Aquaphile™ Coelenterazine h, lyophilized solid
Catalog No. 501964120
Encompass_Preferred
Aquaphile coelenterazine native is a bioluminescence substrate in a special formulation format that is readily dissolved in water or PBS buffer for in vivo use.Properties: Yellow solid soluble in water or PBS buffer; Store at -20°C and protect from light; C26H21N3O3; MW: 423; [55779-48-1].
Catalog No. 50-196-4120 Supplier Biotium Supplier No. 10127
Aquaphile coelenterazine native is a bioluminescence substrate in a special formulation format that is readily dissolved in water or PBS buffer for in vivo use.Properties: Yellow solid soluble in water or PBS buffer; Store at -20°C and protect from light; C26H21N3O3; MW: 423; [55779-48-1].