Learn More
Biotium Aquaphile™ Coelenterazine Native, lyophilized solid
Supplier:  Biotium 1012650UG
Encompass_Preferred
Aquaphile coelenterazine native is a bioluminescence substrate in a special formulation format that is readily dissolved in water or PBS buffer for in vivo use.Properties: Yellow solid soluble in water or PBS buffer; Store at -20°C and protect from light; C26H21N3O3; MW: 423; [55779-48-1].
Catalog No. 50-196-4119