Learn More
Biotium Aquaphile™ Coelenterazine Native, lyophilized solid
Catalog No. NC1283078
Encompass_Preferred
Aquaphile coelenterazine native is a bioluminescence substrate in a special formulation format that is readily dissolved in water or PBS buffer for in vivo use.Properties: Yellow solid soluble in water or PBS buffer; Store at -20°C and protect from light; C26H21N3O3; MW: 423; [55779-48-1]
Catalog No. NC1283078 Supplier Biotium Supplier No. 10126
Aquaphile coelenterazine native is a bioluminescence substrate in a special formulation format that is readily dissolved in water or PBS buffer for in vivo use.Properties: Yellow solid soluble in water or PBS buffer; Store at -20°C and protect from light; C26H21N3O3; MW: 423; [55779-48-1]