Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ AquaPhoenix™ Adenosine Triphosphate (ATP)
SCI_ED

Catalog No. s25123 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25123 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25123
Only null left
Add to Cart
Add to Cart

Powder

  • Quantity: 1g
  • Form: Powder
  • Suitable for use in many molecular biology applications

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Format Solid
Molecular Weight (g/mol) 551.18
Content And Storage Store in a cool location. Keep away from food and beverages. Protect from freezing and physical damage. Provide ventilation for containers. Keep container tightly sealed. Store away from incompatible materials.
Quantity 1 g
Color White
For Use With (Application) Molecular biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.