Learn More
Aqua Solutions AquaPhoenix Scientific Phenolphthalein Indicator Solution, 1.0% w/v, in Isopropanol, 1 L
SDP
Catalog No. NC3994875
Encompass_Preferred
Phenolphthalein Indicator Solution, 1.0% w/v in Isopropanol, is a chemical solution designed for use as an indicator. It is classified as highly flammable and carries several hazard classifications related to health and safety.
  • Highly flammable liquid and vapor.
  • Causes serious eye irritation.
  • Suspected of causing genetic defects.
  • May cause cancer.
  • Suspected of damaging fertility or the unborn child.
  • May cause drowsiness or dizziness.
  • Contains Propan-2-ol (43.5%), Phenolphthalein (1.1%), and Water (55.4%).
Catalog No. NC3994875 Supplier Aqua Solutions Supplier No. 66631L
Phenolphthalein Indicator Solution, 1.0% w/v in Isopropanol, is a chemical solution designed for use as an indicator. It is classified as highly flammable and carries several hazard classifications related to health and safety.
  • Highly flammable liquid and vapor.
  • Causes serious eye irritation.
  • Suspected of causing genetic defects.
  • May cause cancer.
  • Suspected of damaging fertility or the unborn child.
  • May cause drowsiness or dizziness.
  • Contains Propan-2-ol (43.5%), Phenolphthalein (1.1%), and Water (55.4%).