Learn More
National Diagnostics AQUAPOR LE GTAC 100 GR
SDP
Catalog No. 5089990018
Encompass
AquaPor LE GTAC, High quality, general purpose agarose. Low boil-over. Low EEO. DNase and RNase free, 100 GR
Catalog No. 50-899-90018 Supplier National Diagnostics Supplier No. EC202100GRM
AquaPor LE GTAC, High quality, general purpose agarose. Low boil-over. Low EEO. DNase and RNase free, 100 GR