Learn More
Perkin Elmer US LLC Brownlee Aquapore RP-300 Column - 30 mm x 4.6 mm I.D., Pkg. 2
Catalog No. NC1934269
Encompass_Preferred
Recommended for the separation of large biomolecules, such as peptides and proteins.
Catalog No. NC1934269 Supplier Perkin Elmer US LLC Supplier No. 07110055
Recommended for the separation of large biomolecules, such as peptides and proteins.