Learn More
BURKLE INC AquaSampler, FEP/PTFE, 35.50 oz. (1050 ml), Lxouter-Ø 42.13x1.81 in. (107cmx46mm)
Catalog No. 5000025968
Encompass_Preferred
Sampler for ground water samples from wells, pits and other sources. Fits all normal well shafts with control openings to Ø 2", without tilting or getting stuck. Ultra pure materials guarantee the highest degree of sample purity. Easy to clean thanks to rounded edges and threads.
Catalog No. 50-000-25968 Supplier BURKLE INC Supplier No. 53081050
Sampler for ground water samples from wells, pits and other sources. Fits all normal well shafts with control openings to Ø 2", without tilting or getting stuck. Ultra pure materials guarantee the highest degree of sample purity. Easy to clean thanks to rounded edges and threads.