Learn More
BURKLE INC AquaSampler, FEP/PTFE, 11.83 oz. (350 ml), Lxouter-Ø 15.75x1.81 in. (40cmx46mm)
Catalog No. 5000025629
Encompass_Preferred
Sampler for ground water samples from wells, pits and other sources. Fits all normal well shafts with control openings to Ø 2", without tilting or getting stuck. Ultra pure materials guarantee the highest degree of sample purity. Easy to clean thanks to rounded edges and threads.
Catalog No. 50-000-25629 Supplier BURKLE INC Supplier No. 53080350
Sampler for ground water samples from wells, pits and other sources. Fits all normal well shafts with control openings to Ø 2", without tilting or getting stuck. Ultra pure materials guarantee the highest degree of sample purity. Easy to clean thanks to rounded edges and threads.