Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Aquasearcher™ Multi-Parameter Analyzer AB33M1

Catalog No. 01922370 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Multiparameter Meter
Multiparameter Meter with Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
01-922-370 Multiparameter Meter with Kit
01-922-371 Multiparameter Meter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-922-370 Supplier OHAUS™ Supplier No. 30589825
Only null left
Add to Cart
Add to Cart

The Aquasearcher™ Multi-Parameter Analyzer AB33M1 is easy-to-use and accurate, capable of measuring up to seven parameters including pH, ORP, conductivity, TDS, salinity, resistivity, and temperature.

  • Features two independent measuring channels which can work simultaneously
  • Equipped with a large 6.5 in. LCD display and a touch keypad
  • Features a 1000 item memory for measurements and calibration trail
  • Provides connectivity and data output capabilities with RS232 and USB interface
  • i-Steward monitors the condition of electrodes to ensure repeatable and consistent measurement accuracy
  • Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
  • Choose between just the AB33M1 meter with electrode holder and a full kit including AB33M1 meter, ST310 electrode, STCON3 probe, electrode holder, 4 pH buffer mini kits (50 mL), standard conductivity solution kits (1413 μS/cm, 50 mL)
TRUSTED_SUSTAINABILITY

Specifications

Outputs RS232, USB Device (Included)
Channels pH, Conductivity
Display Type LCD
IP Rating None
Measurement Modes Auto-stop, Continuous
Range (Conductivity) 0.01 μS/cm to 500 mS/cm
Range (mV) -2000 to 2000 mV
Range (pH) -2 to 20
Range (Resistivity) 2 to 100 MΩ-cm
Range (Salinity) 0 to 100 psu
Range (TDS) 0.1 mg/L to 199.9 g/L
Resolution (ISE) 0.01 pH
Resolution (pH) 0.1/0.01 pH
Temperature Range (Metric) -5°C to +110°C
Includes AB33M1 Meter, ST310 Electrode, STCON3 Probe, Electrode Holder, 4 pH Buffer Mini Kits (50 mL), Standard Conductivity Solution Kits (1413 μS/cm, 50 mL)
Product Type Multiparameter Meter with Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.