Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Aquasearcher™ pH Meter AB23PH
SCI_ED

Catalog No. S05302 Shop All OHAUS Products
Change view
Click to view available options
Type:
pH Meter with Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S05302 pH Meter with Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05302 Supplier OHAUS™ Supplier No. 30589821/EMD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Aquasearcher™ pH Meter AB23PH is a simple to use benchtop meter for measuring pH and oxidation-reduction potential (ORP). An intuitive design and auto-buffer recognition make measuring pH straightforward and easy.

  • Equipped with a large 5 in. LCD display and six instructional menu buttons
  • Auto buffer recognition and 3-point calibration capability ensures a reliable calibration result
  • Features a 99 item memory for measurements and last calibration
  • Choose between just the AB23PH meter with electrode holder and a full kit including AB23PH meter, ST320 pH electrode, and four pH Buffer Mini Kits (50 mL)
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±1
Accuracy (pH) ±0.01
Data Management Details Datalog for Measurements and Calibration
Includes AB23PH Meter, Electrode Holder, ST320 pH Electrode, 4 pH Buffer Mini Kits (50 mL)
Measurement Modes Auto-stop, Continuous
Calibration Linear
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Calibration Points Up to 3
Temperature Range (Metric) 0.0°C to 100.0°C
Display Type LCD
Backlight Option Yes
Data Points 99 Sets
Log Function Type Automatic Data Logging
Type pH Meter with Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.