Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Aquasearcher™ pH Meter AB33PH

Catalog No. 01922369 Shop All OHAUS Products
Change view
Click to view available options
Type:
Meter with pH Kit
pH Meter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-922-369 pH Meter
01-922-368 Meter with pH Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-922-369 Supplier OHAUS™ Supplier No. 30589826
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Aquasearcher™ pH Meter AB33PH is a highly reliable and user-friendly pH benchtop meter with electrode (ST310) for standard laboratory applications. Designed to measure pH and oxidation-reduction potential with temperature measurements.

  • Equipped with a large 6.5 in. LCD display and a touch keypad
  • Features a 1000 item memory for pH measurements and calibration trail
  • Provides connectivity and data output capabilities with RS232 and USB interface
  • i-Steward monitors the condition of electrodes to ensure repeatable and consistent measurement accuracy
  • Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
  • Auto endpoint mode and auto buffer recognition makes calibration easy
  • Choose between just the AB33PH meter with electrode holder and a full kit including AB33PH meter, ST310 electrode, electrode holder, 4 pH buffer mini kits (50 mL)
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Temperature) ±0.5°C
Accuracy (pH) ±0.01
Data Management Details Datalog with Time and Date Stamp
Accuracy (mV) ±1
Outputs USB, RS-232 (PC), RS-232 (Printer)
Includes AB33PH Meter, Electrode Holder
Measurement Modes Auto-stop, Continuous
Calibration Linear
Range (pH) -2 to 16
Resolution (pH) 0.01, 0.1
Range (mV) ±2000.0
Calibration Points Up to 3
Temperature Range (Metric) -5°C to +110°C
Display Type LCD, 6.5 in. Segment and Dot Matrix
Backlight Option LCD
Data Points 1000 Sets
Log Function Type 1000 Measurement, Last Calibration
Type pH Meter
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.