Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Aquasearcher™ pH Meter AB41PH
SCI_ED

Catalog No. S05294 Shop All OHAUS Products
Change view
Click to view available options
Type:
pH Meter with Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S05294 pH Meter with Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05294 Supplier OHAUS™ Supplier No. 30589831/EMD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Aquasearcher™ pH Meter AB41PH is an advanced, research-grade benchtop pH meter with electrode, offering accurate, repeatable results with selectable resolution and an intelligent i-Steward system.

  • Equipped with a large 6.5 in. LCD display and a touch keypad
  • Features a 1000 item memory and password management for GLP documentation
  • Provides connectivity and data output capabilities with RS232 and USB interface
  • i-Steward ensures repeatable and consistent measurement accuracy
  • Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
  • Choose between just the AB41PH meter with electrode holder and a full kit including AB41PH meter, ST410 electrode, STTEMP probe, electrode holder, AS20 magnetic stirrer, and 4 pH buffer mini kits (50 mL)
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Temperature) ±0.3°C
Data Management Details Datalog with Time and Date Stamp
Accuracy (mV) ±0.3
Accuracy (pH) ±0.002
Outputs USB, RS-232 (PC), RS-232 (Printer)
Includes AB41PH Meter, ST410 Electrode, STTEMP Probe, Electrode Holder, AS20 Magnetic Stirrer, 4 pH Buffer Mini Kits (50 mL)
Measurement Modes Auto-stop, Timed, Continuous
Calibration Offset
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Range (mV) ±2000
Calibration Points Up to 5
Temperature Range (Metric) -10°C to +125°C
Display Type LCD, 6.5 in. Segment and Dot Matrix
Backlight Option LCD
Data Points 1000 Sets
Log Function Type 1000 Measurement, 10 Calibration
Type pH Meter with Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.