Learn More
StatLab AquaSlip Mounting Medium
SDP
Catalog No. 21033449
Change view
Click to view available options
Volume (Metric):
30 mL
Catalog No. Volume (Metric)
21-033-449 30 mL
1 options
Catalog No. 21-033-449 Supplier StatLab Supplier No. MMC0619
Add to Cart
Add to Cart
An aqueous mounting medium which prevents stained tissue chromogens from fading while allowing permanent storage of stained slides
  • With its high refractive index, AquaSlip does not require coverslipping and is compatible with chromogens such as AEC, DAB, Fast Red, BCIP/NBT, and BCIP/INT
  • Prevents stained tissue chromogens from fading
  • 30ml
Specifications
Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 30 mL